Assay Details
| Gene Symbol: Akp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007432.2 | NP_031458.2 | 10 - 11 | 1300 | 107 | |
| GenBank mRNA | BC115745.1 | - | 10 - 11 | 1299 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129621 | BC145627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129621 | NM_007432.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129621 | BC115745.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860738 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511881 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102403 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129621 | BC145627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129621 | NM_007432.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129621 | BC115745.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P299344 | BC145627.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P299344 | NM_007432.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P299344 | BC115745.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20415870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102403 | - | - |
| Whole Mouse Genome G4122A | A_51_P299344 | BC145627.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299344 | NM_007432.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299344 | BC115745.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140586 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139532 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102403 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00577784 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00577803 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00577797 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00577798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20415904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01123071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00577798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00345856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00495639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00897936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00127270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00127269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00488756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00495635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00577783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20415898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00243717 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10348140 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348138 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348137 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348141 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348139 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348132 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348134 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348135 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348136 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348142 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450572_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10348131 | - | - |
| Mouse Genome 430 2.0 Array | 1450572_at | - | - |
| Mouse Genome 430 2.0 Array | 1450572_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11648_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11648_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100006739.mm.1 | NM_007432.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100006739.mm.1 | NM_007432.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100006739.mm.1 | TC0100000806.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450572_PM_at | - | - |
| Mouse Expression Set 430 | 1450572_at | - | - |
| Murine Genome U74v2 Set | 102147_at | - | - |
| Murine Genome U74v2 Set | 102147_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450572_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450572_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450572_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450572_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 699894 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6420301 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6420301 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006420301 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2698889 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2698889 | - | - |
| mouse-6 v1.1 (Illumina) | scl14563.10.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6420301 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl14563.10.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420301 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2698889 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2698889 | - | - |

Akp3
Genomic Map