Assay Details
| Gene Symbol: Akp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007432.2 | NP_031458.2 | 6 - 7 | 782 | 75 | |
| GenBank mRNA | BC115745.1 | - | 6 - 7 | 781 | 75 | |
| BC145627.1 | - | 4 - 5 | 979 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akp3
Genomic Map