| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200002453.mm.1 |
NM_178767.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002453.mm.1 |
NM_178767.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002453.mm.1 |
TC1200000334.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002460.mm.1 |
NM_178767.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002460.mm.1 |
NM_178767.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002460.mm.1 |
TC1200000334.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002455.mm.1 |
NM_178767.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002455.mm.1 |
NM_178767.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002455.mm.1 |
TC1200000334.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002456.mm.1 |
NM_178767.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002456.mm.1 |
NM_178767.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002456.mm.1 |
TC1200000334.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002457.mm.1 |
NM_178767.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002457.mm.1 |
NM_178767.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002457.mm.1 |
TC1200000334.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002454.mm.1 |
NM_178767.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002454.mm.1 |
NM_178767.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002454.mm.1 |
TC1200000334.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002458.mm.1 |
NM_178767.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002458.mm.1 |
NM_178767.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002458.mm.1 |
TC1200000334.mm.1 |
MTA TC |