Assay Details
| Gene Symbol: Agmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178767.5 | NP_848882.2 | 8 - 9 | 1221 | 77 | |
| GenBank mRNA | AK039191.1 | - | 8 - 9 | 792 | 77 | |
| AK040702.1 | - | 8 - 9 | 1222 | 77 | ||
| AK163155.1 | - | 8 - 9 | 1002 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agmo
Genomic Map