Assay Details
| Gene Symbol: Dars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172644.3 | NP_766232.1 | 10 - 11 | 1535 | 88 | |
| GenBank mRNA | AK030952.1 | - | 10 - 11 | 1524 | 88 | |
| AK033634.1 | - | 9 - 10 | 2358 | 88 | ||
| AK037337.1 | - | 10 - 11 | 1554 | 88 | ||
| AK048454.1 | - | 10 - 11 | 1512 | 88 | ||
| AK138809.1 | - | 10 - 11 | 1536 | 88 | ||
| AK154133.1 | - | 10 - 11 | 1563 | 88 | ||
| AK154348.1 | - | 10 - 11 | 1563 | 88 | ||
| AK164885.1 | - | 10 - 11 | 1505 | 88 | ||
| BC046429.1 | - | 3 - 4 | 300 | 88 | ||
| BC057943.1 | - | 10 - 11 | 1559 | 88 | ||
| BC076606.1 | - | 10 - 11 | 1527 | 30625515 | 88 | |
| BC089191.1 | - | 10 - 11 | 1509 | 30473852 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dars2
Genomic Map