Assay Details
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172644.3 | NP_766232.1 | 12 - 13 | 1698 | 109 | |
GenBank mRNA | AK030952.1 | - | 12 - 13 | 1687 | 109 | |
AK033634.1 | - | 11 - 12 | 2521 | 109 | ||
AK037337.1 | - | 12 - 13 | 1717 | 109 | ||
AK048454.1 | - | 12 - 13 | 1675 | 109 | ||
AK138809.1 | - | 12 - 13 | 1699 | 109 | ||
AK154133.1 | - | 12 - 13 | 1726 | 109 | ||
AK154348.1 | - | 12 - 13 | 110 | |||
AK164885.1 | - | 12 - 13 | 1668 | 109 | ||
AK206226.1 | - | 2 - 3 | 122 | 109 | ||
AK210088.1 | - | 3 - 4 | 193 | 109 | ||
BC046429.1 | - | 5 - 6 | 463 | 109 | ||
BC076606.1 | - | 12 - 13 | 1690 | 30625515 | 109 | |
BC089191.1 | - | 12 - 13 | 1672 | 30473852 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
