Assay Details
| Gene Symbol: Dars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172644.3 | NP_766232.1 | 5 - 6 | 999 | 98 | |
| GenBank mRNA | AK030952.1 | - | 5 - 6 | 988 | 98 | |
| AK033634.1 | - | 4 - 5 | 1822 | 98 | ||
| AK037337.1 | - | 5 - 6 | 1018 | 98 | ||
| AK048454.1 | - | 5 - 6 | 976 | 98 | ||
| AK138809.1 | - | 5 - 6 | 1000 | 98 | ||
| AK154133.1 | - | 5 - 6 | 1028 | 98 | ||
| AK154348.1 | - | 5 - 6 | 1028 | 98 | ||
| AK184189.1 | - | 2 - 3 | 187 | 98 | ||
| AK184192.1 | - | 2 - 3 | 187 | 98 | ||
| BC057943.1 | - | 5 - 6 | 1023 | 98 | ||
| BC076606.1 | - | 5 - 6 | 991 | 30625515 | 98 | |
| BC089191.1 | - | 5 - 6 | 973 | 30473852 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dars2
Genomic Map