Assay Details
| Gene Symbol: Csad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144942.4 | NP_659191.1 | 2 - 3 | 295 | 80 | |
| GenBank mRNA | AK005015.1 | - | 2 - 3 | 296 | 80 | |
| AY033912.1 | - | 2 - 3 | 212 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csad
Genomic Map