Assay Details
Gene Symbol: Ddx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017397.3 | NP_059093.3 | 8 - 9 | 1250 | 67 | |
GenBank mRNA | AF217000.1 | - | 8 - 9 | 1125 | 67 | |
AF220454.1 | - | 8 - 9 | 1253 | 67 | ||
AJ250027.1 | - | 8 - 9 | 1125 | 67 | ||
AK154518.1 | - | 8 - 9 | 1129 | 67 | ||
BC125471.1 | - | 8 - 9 | 1250 | 40129986 | 67 | |
BC137721.1 | - | 8 - 9 | 1250 | 8860740 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
