Assay Details
| Gene Symbol: Fam19a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252310.1 | NP_001239239.1 | 2 - 3 | 452 | 95 | |
| NM_134096.2 | NP_598857.1 | 2 - 3 | 404 | 95 | ||
| GenBank mRNA | AK090194.1 | - | 2 - 3 | 278 | 95 | |
| AK090307.1 | - | 2 - 3 | 454 | 95 | ||
| AY325124.1 | - | 2 - 3 | 405 | 95 | ||
| BC015306.1 | - | 2 - 3 | 405 | 4223181 | 95 | |
| BI986174.1 | - | 1 - 2 | 84 | 95 | ||
| BU057253.1 | - | 2 - 3 | 408 | 95 | ||
| BY733542.1 | - | 2 - 3 | 454 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam19a5
Genomic Map