Assay Details
| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011479.3 | NP_035609.1 | 8 - 9 | 1223 | 63 | |
| GenBank mRNA | AK044917.1 | - | 8 - 9 | 1224 | 63 | |
| AK057644.1 | - | 8 - 9 | 1259 | 63 | ||
| AK080181.1 | - | 2 - 3 | 1421 | 63 | ||
| AK089019.1 | - | 8 - 9 | 1217 | 63 | ||
| AK133728.1 | - | 2 - 3 | 284 | 63 | ||
| AK159346.1 | - | 8 - 9 | 1224 | 63 | ||
| AK177584.1 | - | 1 - 2 | 58 | 63 | ||
| BC003227.1 | - | 8 - 9 | 1256 | 3483158 | 63 | |
| U27455.1 | - | 8 - 9 | 1191 | 63 | ||
| X95642.1 | - | 8 - 9 | 1194 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sptlc2
Genomic Map