Assay Details
| Gene Symbol: Ism2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290302.1 | NP_001277231.1 | 3 - 4 | 740 | 109 | |
| GenBank mRNA | BU054611.1 | - | 2 - 3 | 545 | 109 | |
More Information
Set Membership: |
|

Ism2
Genomic Map