Assay Details
| Gene Symbol: Isl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021459.4 | NP_067434.3 | 3 - 4 | 748 | 105 | |
| GenBank mRNA | AK090263.1 | - | 3 - 4 | 749 | 105 | |
| BC132263.1 | - | 3 - 4 | 674 | 40130540 | 105 | |
| BC132609.1 | - | 3 - 4 | 674 | 40130886 | 105 | |
| CF725499.1 | - | 2 - 3 | 383 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Isl1
Genomic Map