Assay Details
| Gene Symbol: Isl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021459.4 | NP_067434.3 | 4 - 5 | 1036 | 65 | |
| GenBank mRNA | AJ132765.1 | - | 4 - 5 | 902 | 65 | |
| AK090263.1 | - | 4 - 5 | 1037 | 65 | ||
| BC132263.1 | - | 4 - 5 | 962 | 40130540 | 65 | |
| BC132609.1 | - | 4 - 5 | 962 | 40130886 | 65 | |
| EU704681.1 | - | 1 - 2 | 94 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Isl1
Genomic Map