Assay Details
Gene Symbol: Upf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026573.2 | NP_080849.1 | 2 - 3 | 355 | 127 | |
GenBank mRNA | AK011606.1 | - | 2 - 3 | 353 | 127 | |
AK019989.1 | - | 2 - 3 | 333 | 127 | ||
AK145431.1 | - | 2 - 3 | 358 | 127 | ||
AK162222.1 | - | 2 - 3 | 351 | 127 | ||
BC058761.1 | - | 2 - 3 | 336 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
