Assay Details
| Gene Symbol: Lhx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008498.2 | NP_032524.1 | 3 - 4 | 2034 | 60 | |
| GenBank mRNA | AK032067.1 | - | 225 | 60 | ||
| AK141086.1 | - | 2036 | 60 | |||
| BC092374.1 | - | 3 - 4 | 2167 | 30363447 | 60 | |
| S68107.1 | - | 6 - 7 | 1566 | 60 | ||
| Z27410.1 | - | 3 - 4 | 1921 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lhx1
Genomic Map