Assay Details
| Gene Symbol: Akap14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033785.2 | NP_001028957.1 | 3 - 4 | 1366 | 113 | |
| GenBank mRNA | AK133120.1 | - | 3 - 4 | 1367 | 113 | |
| BC147265.1 | - | 3 - 4 | 1366 | 9056322 | 113 | |
| BC147494.1 | - | 3 - 4 | 1366 | 9056315 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap14
Genomic Map