Assay Details
| Gene Symbol: Akap14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033785.2 | NP_001028957.1 | 5 - 6 | 1606 | 89 | |
| GenBank mRNA | AK133120.1 | - | 5 - 6 | 1607 | 89 | |
| BC147265.1 | - | 5 - 6 | 1606 | 9056322 | 89 | |
| BC147494.1 | - | 5 - 6 | 1606 | 9056315 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P639751 | BC147494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P639751 | NM_001033785.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P639751 | AK133120.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P639751 | BC147265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P639751 | BC147494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P639751 | NM_001033785.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P639751 | AK133120.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P639751 | BC147265.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496587 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265682 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502828 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216155 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121789 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149834 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P639751 | BC147494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P639751 | NM_001033785.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P639751 | AK133120.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P639751 | BC147265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P639751 | BC147494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P639751 | NM_001033785.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P639751 | AK133120.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P639751 | BC147265.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03656383 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149834 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09265648 | - | - |
| Whole Mouse Genome G4122A | A_52_P639751 | BC147494.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P639751 | NM_001033785.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P639751 | AK133120.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P639751 | BC147265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P639751 | BC147494.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P639751 | NM_001033785.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P639751 | AK133120.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P639751 | BC147265.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32321964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14595192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14954058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15033331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14693762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14617207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15206956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15100426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14986510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14779977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15033337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14789414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14932674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14986507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32321992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322044 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10604095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604096 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457541_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10604094 | - | - |
| Mouse Genome 430 2.0 Array | 1457541_at | - | - |
| Mouse Genome 430 2.0 Array | 1457541_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 434756_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 434756_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00013105.mm.1 | NM_001033785.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013105.mm.1 | NM_001033785.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013105.mm.1 | TC0X00002060.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013106.mm.1 | NM_001033785.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013106.mm.1 | NM_001033785.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013106.mm.1 | TC0X00002060.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457541_PM_at | - | - |
| Mouse Expression Set 430 | 1457541_at | - | - |
| Murine Genome U74v2 Set | 167385_at | - | - |
| Murine Genome U74v2 Set | 167385_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457541_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457541_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2780362 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2780362 | - | - |

Akap14
Genomic Map