Assay Details
| Gene Symbol: Adamts6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081020.1 | NP_001074489.1 | 3 - 4 | 866 | 169 | |
| GenBank mRNA | AK044528.1 | - | 3 - 4 | 869 | 169 | |
| BC172116.1 | - | 3 - 4 | 764 | 169 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adamts6
Genomic Map