Assay Details
| Gene Symbol: Adamts6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081020.1 | NP_001074489.1 | 6 - 7 | 1337 | 68 | |
| GenBank mRNA | AK030315.1 | - | 6 - 7 | 1490 | 68 | |
| AK044528.1 | - | 6 - 7 | 1340 | 68 | ||
| BC172116.1 | - | 6 - 7 | 1240 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adamts6
Genomic Map