Assay Details
| Gene Symbol: Slc13a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022411.3 | NP_071856.1 | 7 - 8 | 1095 | 60 | |
| GenBank mRNA | AF201903.1 | - | 7 - 8 | 1095 | 60 | |
| AK140972.1 | - | 7 - 8 | 1146 | 60 | ||
| BC013493.1 | - | 7 - 8 | 1120 | 4218337 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc13a2
Genomic Map