Assay Details
| Gene Symbol: Mpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010824.2 | NP_034954.2 | 10 - 11 | 120 | ||
| GenBank mRNA | AK160229.1 | - | 9 - 10 | 120 | ||
| AK220229.1 | - | 9 - 10 | 1613 | 120 | ||
| AY500847.1 | - | 10 - 11 | 1594 | 120 | ||
| BC053912.1 | - | 9 - 10 | 30044601 | 120 | ||
| X15313.1 | - | 10 - 11 | 2050 | 120 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mpo
Genomic Map