Assay Details
| Gene Symbol: Bzrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172449.2 | NP_766037.2 | 14 - 15 | 2725 | 64 | |
| GenBank mRNA | CB249809.1 | - | 4 - 5 | 465 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bzrap1
Genomic Map