Assay Details
| Gene Symbol: Bzrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172449.2 | NP_766037.2 | 16 - 17 | 2952 | 82 | |
| GenBank mRNA | CD348883.1 | - | 2 - 3 | 244 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bzrap1
Genomic Map