| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100012684.mm.1 |
NM_172449.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012684.mm.1 |
NM_172449.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012684.mm.1 |
TC1100001352.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012685.mm.1 |
NM_172449.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012685.mm.1 |
NM_172449.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012685.mm.1 |
TC1100001352.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012687.mm.1 |
NM_172449.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012687.mm.1 |
NM_172449.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012687.mm.1 |
TC1100001352.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012686.mm.1 |
NM_172449.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012686.mm.1 |
NM_172449.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012686.mm.1 |
TC1100001352.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012688.mm.1 |
NM_172449.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012688.mm.1 |
NM_172449.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012688.mm.1 |
TC1100001352.mm.1 |
MTA TC |