Assay Details
| Gene Symbol: Bzrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172449.2 | NP_766037.2 | 22 - 23 | 5568 | 134 | |
| GenBank mRNA | AK051895.1 | - | 1 - 2 | 1517 | 134 | |
| BC066146.1 | - | 1 - 2 | 579 | 30470084 | 134 | |
| EL605892.1 | - | 2 - 3 | 134 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC066146.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bzrap1
Genomic Map