Assay Details
| Gene Symbol: Bzrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172449.2 | NP_766037.2 | 30 - 31 | 6371 | 113 | |
| GenBank mRNA | AK122329.1 | - | 7 - 8 | 1452 | 113 | |
| BC055691.1 | - | 10 - 11 | 1122 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bzrap1
Genomic Map