Assay Details
| Gene Symbol: Pdgfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011057.3 | NP_035187.2 | 2 - 3 | 968 | 90 | |
| GenBank mRNA | AF162784.1 | - | 2 - 3 | 158 | 90 | |
| AK087840.1 | - | 2 - 3 | 1143 | 90 | ||
| BC023427.1 | - | 1 - 2 | 92 | 90 | ||
| BC053430.1 | - | 2 - 3 | 968 | 6330609 | 90 | |
| BC064056.1 | - | 2 - 3 | 635 | 1380782 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfb
Genomic Map