Assay Details
| Gene Symbol: Mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044773.1 | - | 2 - 3 | 802 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044773.1 transcripts.
Set Membership: |
|

Mief1
Genomic Map