Assay Details
| Gene Symbol: Kpnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008379.3 | NP_032405.3 | 18 - 19 | 2548 | 140 | |
| GenBank mRNA | AK077709.1 | - | 18 - 19 | 2539 | 140 | |
| AK147175.1 | - | 18 - 19 | 2567 | 140 | ||
| AK169171.1 | - | 18 - 19 | 2515 | 140 | ||
| BC052438.1 | - | 18 - 19 | 2303 | 6405935 | 140 | |
| BC052711.1 | - | 18 - 19 | 2336 | 6835239 | 140 | |
| BC055115.1 | - | 18 - 19 | 2533 | 6518750 | 140 | |
| D45836.1 | - | 18 - 19 | 2439 | 140 | ||
| D67015.1 | - | 18 - 19 | 2276 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kpnb1
Genomic Map