Assay Details
| Gene Symbol: Kpnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008379.3 | NP_032405.3 | 20 - 21 | 2771 | 100 | |
| GenBank mRNA | AK077709.1 | - | 20 - 21 | 2762 | 100 | |
| AK147175.1 | - | 20 - 21 | 2790 | 100 | ||
| AK169171.1 | - | 20 - 21 | 2738 | 100 | ||
| BC052438.1 | - | 20 - 21 | 2526 | 6405935 | 100 | |
| BC052711.1 | - | 20 - 21 | 2559 | 6835239 | 100 | |
| BC055115.1 | - | 20 - 21 | 2756 | 6518750 | 100 | |
| D45836.1 | - | 20 - 21 | 2662 | 100 | ||
| D67015.1 | - | 20 - 21 | 2499 | 100 | ||
More Information
Set Membership: |
|

Kpnb1
Genomic Map