Assay Details
| Gene Symbol: Uhrf1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080769.1 | NP_001074238.1 | 11 - 12 | 1668 | 76 | |
| GenBank mRNA | AK144200.1 | - | 6 - 7 | 964 | 76 | |
| AK220293.1 | - | 11 - 12 | 1668 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Uhrf1bp1
Genomic Map