Assay Details
| Gene Symbol: Uhrf1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080769.1 | NP_001074238.1 | 19 - 20 | 4307 | 64 | |
| GenBank mRNA | AK089790.1 | - | 19 - 20 | 4335 | 64 | |
| AK144200.1 | - | 14 - 15 | 3603 | 64 | ||
| AK165438.1 | - | 1 - 2 | 45 | 64 | ||
| AK220293.1 | - | 19 - 20 | 4307 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Uhrf1bp1
Genomic Map