| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700003935.mm.1 |
NM_001080769.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003935.mm.1 |
NM_001080769.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003935.mm.1 |
TC1700000469.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700003930.mm.1 |
NM_001080769.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003930.mm.1 |
NM_001080769.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003930.mm.1 |
TC1700000469.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700003933.mm.1 |
NM_001080769.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003933.mm.1 |
NM_001080769.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003933.mm.1 |
TC1700000469.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700003932.mm.1 |
NM_001080769.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003932.mm.1 |
NM_001080769.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003932.mm.1 |
TC1700000469.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700003934.mm.1 |
NM_001080769.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003934.mm.1 |
NM_001080769.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700003934.mm.1 |
TC1700000469.mm.1 |
MTA TC |