Assay Details
| Gene Symbol: Uhrf1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080769.1 | NP_001074238.1 | 5 - 6 | 770 | 70 | |
| GenBank mRNA | AK220293.1 | - | 5 - 6 | 770 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Uhrf1bp1
Genomic Map