Assay Details
| Gene Symbol: C1qtnf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204134.1 | NP_001191063.1 | 2 - 3 | 516 | 128 | |
| NM_030888.4 | NP_112150.1 | 2 - 3 | 322 | 128 | ||
| GenBank mRNA | AF246265.1 | - | 2 - 3 | 299 | 128 | |
| AK029352.1 | - | 2 - 3 | 299 | 128 | ||
| AK075867.1 | - | 2 - 3 | 299 | 128 | ||
| AK081704.1 | - | 2 - 3 | 299 | 128 | ||
| AK128898.1 | - | 2 - 3 | 322 | 128 | ||
| BC019636.1 | - | 2 - 3 | 525 | 128 | ||
| BC055047.1 | - | 2 - 3 | 284 | 6486507 | 128 | |
| BC055859.1 | - | 2 - 3 | 263 | 4238617 | 128 | |
| DQ002396.1 | - | 2 - 3 | 200 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C1qtnf3
Genomic Map