Assay Details
| Gene Symbol: Ttc23l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029430.1 | NP_083706.1 | 1 - 2 | 77 | 65 | |
| GenBank mRNA | AK015027.1 | - | 1 - 2 | 49 | 65 | |
| BC132384.1 | - | 1 - 2 | 77 | 65 | ||
| BC145852.1 | - | 1 - 2 | 77 | 40131155 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015027.1 transcripts.
Set Membership: |
|

Ttc23l
Genomic Map