Assay Details
| Gene Symbol: Ttc23l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029430.1 | NP_083706.1 | 9 - 10 | 1219 | 121 | |
| GenBank mRNA | AK015027.1 | - | 9 - 10 | 1191 | 121 | |
| BC132384.1 | - | 9 - 10 | 1219 | 121 | ||
| BC145852.1 | - | 9 - 10 | 1219 | 40131155 | 121 | |
More Information
Set Membership: |
|

Ttc23l
Genomic Map