Assay Details
| Gene Symbol: Cdpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164625.1 | NP_001158097.1 | 3 - 4 | 374 | 117 | |
| NM_001164626.1 | NP_001158098.1 | 2 - 3 | 168 | 117 | ||
| NM_197998.2 | NP_932115.2 | 3 - 4 | 315 | 117 | ||
| GenBank mRNA | BC025858.1 | - | 3 - 4 | 277 | 5007877 | 117 |
| BQ550379.1 | - | 3 - 4 | 333 | 117 | ||
| BY081241.1 | - | 3 - 4 | 277 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403117 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403119 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403116 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403118 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | NM_001164625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | NM_001164626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | NM_197998.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | AI849737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | AK008763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | NM_001164625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | NM_001164626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | NM_197998.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | AI849737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P450835 | AK008763.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954244 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154253 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | NM_001164625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | NM_001164626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | NM_197998.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | AI849737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | AK008763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | NM_001164625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | NM_001164626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | NM_197998.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | AI849737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P450835 | AK008763.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30403121 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03089868 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154253 | - | - |
| Whole Mouse Genome G4122A | A_52_P450835 | NM_001164625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450835 | NM_001164626.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450835 | NM_197998.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450835 | AI849737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450835 | AK008763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450835 | NM_001164625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450835 | NM_001164626.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450835 | NM_197998.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450835 | AI849737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450835 | AK008763.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119780 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154253 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12044176 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12044192 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12044179 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12044181 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12044183 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403116 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403119 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403117 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403118 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30403121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30403170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12044176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12044179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12044181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12308323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12308350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12205917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12044190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11968661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12044192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12308341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30403128 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10431223 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431224 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431225 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431222 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451409_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10431220 | - | - |
| Mouse Genome 430 2.0 Array | 1451409_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451409_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72355_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72355_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500015932.mm.1 | NM_001164625.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015932.mm.1 | NM_001164625.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015932.mm.1 | NM_001164626.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015932.mm.1 | NM_001164626.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015932.mm.1 | NM_197998.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015932.mm.1 | NM_197998.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015932.mm.1 | TC1500002007.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015931.mm.1 | NM_001164625.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015931.mm.1 | NM_001164625.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015931.mm.1 | NM_001164626.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015931.mm.1 | NM_001164626.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015931.mm.1 | NM_197998.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015931.mm.1 | NM_197998.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015931.mm.1 | TC1500002007.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015929.mm.1 | NM_001164625.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015929.mm.1 | NM_001164625.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015929.mm.1 | NM_001164626.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015929.mm.1 | NM_001164626.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015929.mm.1 | NM_197998.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015929.mm.1 | NM_197998.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015929.mm.1 | TC1500002007.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015926.mm.1 | NM_001164625.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015926.mm.1 | NM_001164625.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015926.mm.1 | NM_001164626.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015926.mm.1 | NM_001164626.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015926.mm.1 | NM_197998.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015926.mm.1 | NM_197998.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015926.mm.1 | TC1500002007.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015925.mm.1 | NM_001164625.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015925.mm.1 | NM_001164625.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015925.mm.1 | NM_001164626.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015925.mm.1 | NM_001164626.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015925.mm.1 | NM_197998.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015925.mm.1 | NM_197998.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015925.mm.1 | TC1500002007.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451409_at | - | - |
| Mouse Expression Set 430 | 1451409_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451409_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451409_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451409_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451409_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 816904 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5550400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5550400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250128 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250128 | - | - |
| mouse-6 v1.1 (Illumina) | scl46876.5.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550400 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46876.5.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550400 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2781118 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2778076 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2781118 | - | - |

Cdpf1
Genomic Map