Assay Details
| Gene Symbol: Tmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198295.2 | NP_938037.2 | 12 - 13 | 1024 | 137 | |
| GenBank mRNA | AK035946.1 | - | 12 - 13 | 972 | 137 | |
| AK042787.1 | - | 12 - 13 | 967 | 137 | ||
| AK048502.1 | - | 12 - 13 | 980 | 137 | ||
| AK050932.1 | - | 12 - 13 | 907 | 137 | ||
| AK129451.1 | - | 12 - 13 | 918 | 137 | ||
| AK140709.1 | - | 12 - 13 | 1025 | 137 | ||
| BC057139.1 | - | 7 - 8 | 492 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmx3
Genomic Map