Assay Details
| Gene Symbol: Tmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198295.2 | NP_938037.2 | 2 - 3 | 268 | 114 | |
| GenBank mRNA | AK035946.1 | - | 2 - 3 | 216 | 114 | |
| AK042787.1 | - | 2 - 3 | 211 | 114 | ||
| AK050932.1 | - | 2 - 3 | 151 | 114 | ||
| AK129451.1 | - | 2 - 3 | 162 | 114 | ||
| AK140709.1 | - | 2 - 3 | 269 | 114 | ||
| AK157865.1 | - | 2 - 3 | 219 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmx3
Genomic Map