Assay Details
| Gene Symbol: Tmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198295.2 | NP_938037.2 | 3 - 4 | 313 | 150 | |
| GenBank mRNA | AK035946.1 | - | 3 - 4 | 261 | 150 | |
| AK042787.1 | - | 3 - 4 | 256 | 150 | ||
| AK048502.1 | - | 3 - 4 | 269 | 150 | ||
| AK050932.1 | - | 3 - 4 | 196 | 150 | ||
| AK129451.1 | - | 3 - 4 | 207 | 150 | ||
| AK140709.1 | - | 3 - 4 | 314 | 150 | ||
| AK157865.1 | - | 3 - 4 | 264 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmx3
Genomic Map