Assay Details
| Gene Symbol: Tmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198295.2 | NP_938037.2 | 5 - 6 | 490 | 79 | |
| GenBank mRNA | AK035946.1 | - | 5 - 6 | 438 | 79 | |
| AK042787.1 | - | 5 - 6 | 433 | 79 | ||
| AK048502.1 | - | 5 - 6 | 446 | 79 | ||
| AK050932.1 | - | 5 - 6 | 373 | 79 | ||
| AK129451.1 | - | 5 - 6 | 384 | 79 | ||
| AK140709.1 | - | 5 - 6 | 491 | 79 | ||
| AK157865.1 | - | 5 - 6 | 441 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmx3
Genomic Map