Assay Details
| Gene Symbol: Tmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198295.2 | NP_938037.2 | 9 - 10 | 817 | 88 | |
| GenBank mRNA | AK035946.1 | - | 9 - 10 | 765 | 88 | |
| AK042787.1 | - | 9 - 10 | 760 | 88 | ||
| AK048502.1 | - | 9 - 10 | 773 | 88 | ||
| AK050932.1 | - | 9 - 10 | 700 | 88 | ||
| AK129451.1 | - | 9 - 10 | 711 | 88 | ||
| AK140709.1 | - | 9 - 10 | 818 | 88 | ||
| AK157865.1 | - | 9 - 10 | 768 | 88 | ||
| BC057139.1 | - | 4 - 5 | 285 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmx3
Genomic Map