Assay Details
| Gene Symbol: Pla2g6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199023.1 | NP_001185952.1 | 6 - 7 | 1671 | 53 | |
| NM_001199024.1 | NP_001185953.1 | 6 - 7 | 1671 | 53 | ||
| NM_001199025.1 | NP_001185954.1 | 7 - 8 | 1176 | 53 | ||
| NM_016915.4 | NP_058611.1 | 6 - 7 | 1217 | 53 | ||
| GenBank mRNA | AF259401.1 | - | 5 - 6 | 951 | 53 | |
| AK144495.1 | - | 7 - 8 | 1050 | 53 | ||
| AK153333.1 | - | 6 - 7 | 1477 | 53 | ||
| BC003487.1 | - | 6 - 7 | 1010 | 2811357 | 53 | |
| BC049778.1 | - | 6 - 7 | 1397 | 53 | ||
| BC052845.1 | - | 6 - 7 | 1042 | 30054573 | 53 | |
| BC057209.1 | - | 6 - 7 | 1264 | 5329147 | 53 | |
| DV647961.1 | - | 2 - 3 | 171 | 53 | ||
| U88624.2 | - | 5 - 6 | 893 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g6
Genomic Map