Assay Details
Gene Symbol: Smg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024262.1 | NP_077224.1 | 2 - 3 | 1924 | 121 | |
GenBank mRNA | AK004714.1 | - | 2 - 3 | 1917 | 121 | |
AK031106.1 | - | 2 - 3 | 1898 | 121 | ||
AK035089.1 | - | 2 - 3 | 891 | 121 | ||
BC002247.1 | - | 2 - 3 | 529 | 121 | ||
BC020005.1 | - | 2 - 3 | 1924 | 3981459 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
