Assay Details
| Gene Symbol: Caskin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027937.2 | NP_082213.2 | 17 - 18 | 1968 | 58 | |
| GenBank mRNA | AK129327.1 | - | 15 - 16 | 1531 | 58 | |
| BC059916.1 | - | 8 - 9 | 800 | 58 | ||
| BC060720.1 | - | 17 - 18 | 1732 | 58 | ||
| BC138443.1 | - | 15 - 16 | 1663 | 8861463 | 58 | |
| BC145281.1 | - | 15 - 16 | 1663 | 9053835 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Caskin1
Genomic Map