Assay Details
| Gene Symbol: Ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144849.1 | NP_659098.1 | 1 - 2 | 389 | 126 | |
| GenBank mRNA | AK050440.1 | - | 1 - 2 | 416 | 126 | |
| AK084500.1 | - | 1 - 2 | 424 | 126 | ||
| AK133785.1 | - | 1 - 2 | 413 | 126 | ||
| AK143625.1 | - | 1 - 2 | 416 | 126 | ||
| BC014743.1 | - | 1 - 2 | 389 | 4503319 | 126 | |
| DQ884878.1 | - | 1 - 2 | 417 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd54
Genomic Map