Assay Details
| Gene Symbol: Ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144849.1 | NP_659098.1 | 4 - 5 | 610 | 58 | |
| GenBank mRNA | AK050440.1 | - | 4 - 5 | 637 | 58 | |
| AK133785.1 | - | 4 - 5 | 634 | 58 | ||
| AK143625.1 | - | 4 - 5 | 637 | 58 | ||
| BC014743.1 | - | 4 - 5 | 610 | 4503319 | 58 | |
| DQ884878.1 | - | 4 - 5 | 638 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd54
Genomic Map