Assay Details
| Gene Symbol: Ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144849.1 | NP_659098.1 | 7 - 8 | 885 | 57 | |
| GenBank mRNA | AK050440.1 | - | 6 - 7 | 1600 | 57 | |
| AK133785.1 | - | 7 - 8 | 909 | 57 | ||
| AK143625.1 | - | 7 - 8 | 912 | 57 | ||
| BC014743.1 | - | 7 - 8 | 885 | 4503319 | 57 | |
| DQ884878.1 | - | 7 - 8 | 913 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364129 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364128 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364126 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364131 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307220 | AK050440.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307220 | AK050440.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334876 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637685 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749821 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15331 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56332 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114113 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307220 | AK050440.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307220 | AK050440.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364137 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114113 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03078125 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08429259 | - | - |
| Whole Mouse Genome G4122A | A_51_P307220 | AK050440.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307220 | AK050440.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118424 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114113 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12311187 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11946934 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12311185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11946935 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11946938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364129 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364126 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364128 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11888704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11984947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12374303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11888708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12451105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12094410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12422729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11946935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12195672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11946930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11888706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364106 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430441 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430442 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430440 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430439 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451182_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438090_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430438 | - | - |
| Mouse Genome 430 2.0 Array | 1438090_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1451182_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1438090_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1451182_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 223690_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 223690_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500014495.mm.1 | NM_144849.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014495.mm.1 | NM_144849.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014495.mm.1 | TC1500001859.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014493.mm.1 | NM_144849.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014493.mm.1 | NM_144849.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014493.mm.1 | TC1500001859.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014494.mm.1 | NM_144849.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014494.mm.1 | NM_144849.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014494.mm.1 | TC1500001859.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451182_PM_s_at | - | - |
| Mouse Expression Set 430 | 1438090_PM_x_at | - | - |
| Mouse Expression Set 430 | 1438090_x_at | - | - |
| Mouse Expression Set 430 | 1451182_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438090_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438090_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451182_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451182_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451182_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438090_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438090_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451182_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 636303 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3440687 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1570463 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3440687 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100050671 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001570463 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440687 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600078 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2722184 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2722184 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600078 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248653 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002525.1_37-S | - | - |
| mouse-6 v1.1 (Illumina) | scl46973.7_42-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1570463 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440687 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100050671 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002525.1_37-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46973.7_42-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100050671 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570463 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440687 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2722184 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2872599 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600078 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2872599 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600078 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2722184 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2579277 | - | - |

Ankrd54
Genomic Map